2-氯乙胺 ≥98.0% (by total nitrogen and titration analysis)

供应商: TCI
Warning

Synonyms: 2-氨基乙基氯

C0851-25G C0851-500G
TCIAC0851-25GEA 0 CNY
TCIAC0851-25G TCIAC0851-500G
2-氯乙胺 ≥98.0% (by total nitrogen and titration analysis)
2-氯乙胺
分子式: C₂H₇Cl₂N
MDL编号: MFCD00012887
CAS号: 870-24-6
EINECS: 212-793-2
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11ee1348877dedc48461d14db79252c6979c38b9&vid=11ee1348877dedc48461d14db79252c6979c38b9&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light red powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 144.0 to 149.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR