2-(4-甲氧苯基)乙胺 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Danger
|
Synonyms:
4-甲氧基苯乙胺, 2-(4-甲氧苯基)乙胺
M0795-25ML
M0795-250ML
TCIAM0795-25MLEA
0
CNY
TCIAM0795-25ML
TCIAM0795-250ML
2-(4-甲氧苯基)乙胺 ≥98.0% (by GC, titration analysis)
2-(4-甲氧苯基)乙胺
分子式:
C₉H₁₃NO 沸点: 138°C |
MDL编号:
MFCD00008192 CAS号: 55-81-2 EINECS: 200-245-5 UN号: 2735 ADR: 8,II |
|
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...