4'-Benzyloxyacetophenone ≥98.0% (by GC)

供应商: TCI

Synonyms: 4-Benzyloxyacetophenone

B3365-1G B3365-5G
TCIAB3365-1GEA 0 CNY
TCIAB3365-1G TCIAB3365-5G
4'-Benzyloxyacetophenone ≥98.0% (by GC)
4'-Benzyloxyacetophenone
分子式: C₁₅H₁₄O₂
分子量: 226.27 g/mol
MDL编号: MFCD00017247
CAS号: 54696-05-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2d09cb22b7653a914709285a04e49c99e4017a0&vid=e2d09cb22b7653a914709285a04e49c99e4017a0&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 91.0 to 95.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR