3-(三氟甲基)溴苄 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: α′-溴-α,α,α-三氟间二甲苯

T1782-25G T1782-5G
TCIAT1782-25GEA 0 CNY
TCIAT1782-25G TCIAT1782-5G
3-(三氟甲基)溴苄 ≥98.0% (by GC)
3-(三氟甲基)溴苄
分子式: C₈H₆BrF₃
分子量: 239.03 g/mol
沸点: 79°C
密度: 1.56
闪点: 88°C
MDL编号: MFCD00000395
CAS号: 402-23-3
EINECS: 206-938-9
UN号: 3265
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e0ab6ef04345750048c43f40d51fc5139b48979&vid=0fb823cb9ef435750c594892ec5a9800973cf2c0&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR