3-甲基苄基溴 ≥98.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
α-溴间二甲苯
B3585-25G
B3585-5G
TCIAB3585-25GEA
0
CNY
TCIAB3585-25G
TCIAB3585-5G
3-甲基苄基溴 ≥98.0% (by GC)
3-甲基苄基溴
分子式:
C₈H₉Br 分子量: 185.06 g/mol 沸点: 98°C 溶点: 18 闪点: 82°C |
MDL编号:
MFCD00000177 CAS号: 620-13-3 EINECS: 210-625-2 UN号: 1701 ADR: 6.1 II Merck Index: 14,10090 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...