α,α′-二溴-间二甲苯 ≥97.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
二溴间二甲苯, 1,3-二(溴甲基)苯
D0215-25G
D0215-5G
TCIAD0215-25GEA
0
CNY
TCIAD0215-25G
TCIAD0215-5G
α,α′-二溴-间二甲苯 ≥97.0% (by GC)
α,α′-二溴-间二甲苯
分子式:
C₈H₈Br₂ 分子量: 263.96 g/mol 沸点: 140°C 存储条件: Ambient |
MDL编号:
MFCD00000178 CAS号: 626-15-3 EINECS: 210-931-6 UN号: 2928 ADR: 6.1 II |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 75.0 to 78.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...