1,3-二苯基-2-丙酮 ≥97.0% (by GC)

供应商: TCI

Synonyms: 1,3-二苯基丙酮, 二苄基甲酮

D0911-25G D0911-500G
TCIAD0911-25GEA 0 CNY
TCIAD0911-25G TCIAD0911-500G
1,3-二苯基-2-丙酮 ≥97.0% (by GC)
1,3-二苯基-2-丙酮
分子式: (C₆H₅)CH₂C(O)CH₂(C₆H₅)
分子量: 210.28 g/mol
沸点: 330
存储条件: Refrigerator
MDL编号: MFCD00004795
CAS号: 102-04-5
EINECS: 203-000-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cdd1a633815c0223978700421f3dfc5f06566cf5&vid=e50d448cc394a36487b427b88a0a9b5057e59988&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White or Colorless to Yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR