4-仲丁基苯酚 ≥98.0% (by GC)

供应商: TCI
Danger

B0733-25G B0733-500G
TCIAB0733-25GEA 0 CNY
TCIAB0733-25G TCIAB0733-500G
4-仲丁基苯酚 ≥98.0% (by GC)
4-仲丁基苯酚
分子式: C₁₀H₁₄O
沸点: 110°C
溶点: 59 °C
密度: 0,9761 g/cm³ (20 °C)
MDL编号: MFCD00002375
CAS号: 99-71-8
EINECS: 202-781-5
UN号: 2430
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef5a5d6dc064ce83c3aa7529d091fa057f004c4b&vid=d3f636b53fe2fd4b4cac8859e4677a5fccd5d882&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 59.0 to 62.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR