对甲基苯甲醚 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: p-Methylanisole

M0149-25ML M0149-500ML
TCIAM0149-25MLEA 0 CNY
TCIAM0149-25ML TCIAM0149-500ML
对甲基苯甲醚 ≥98.0% (by GC)
对甲基苯甲醚
分子式: C₈H₁₀O
分子量: 122.17 g/mol
沸点: 175°C
溶点: -36
闪点: 59°C
存储条件: Ambient
MDL编号: MFCD00008413
CAS号: 104-93-8
EINECS: 203-253-7
UN号: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5998ff110ef1366b10462720b849d5801bd231c8&vid=3bab89bdf1b41b22f83270315876f9600fc32158&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR