间甲基苯甲醚 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
1-Methoxy-3-methylbenzene
M0151-25ML
M0151-500ML
TCIAM0151-25MLEA
0
CNY
TCIAM0151-25ML
TCIAM0151-500ML
间甲基苯甲醚 ≥98.0% (by GC)
间甲基苯甲醚
分子式:
C₈H₁₀O 分子量: 122.17 g/mol 沸点: 177°C 溶点: -47 闪点: 54°C 存储条件: Ambient |
MDL编号:
MFCD00008395 CAS号: 100-84-5 EINECS: 202-893-4 UN号: 3271 ADR: 3 III |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...