2-Ethylanisole ≥98.0% (by GC)
供应商: TCI
Synonyms:
1-Ethyl-2-methoxybenzene, 2-Ethylmethoxybenzene
E0509-25G
E0509-5G
TCIAE0509-25GEA
0
CNY
TCIAE0509-25G
TCIAE0509-5G
2-Ethylanisole ≥98.0% (by GC)
2-Ethylanisole
分子式:
C₉H₁₂O |
MDL编号:
MFCD00142909 CAS号: 14804-32-1 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...