2-氟-4-碘甲苯 ≥95.0% (by GC)

供应商: TCI
Warning

Synonyms: 1-Fluoro-5-iodo-2-methylbenzene, 2-Fluoro-4-iodo-1-methylbenzene

F0720-25G F0720-5G
TCIAF0720-25GEA 0 CNY
TCIAF0720-25G TCIAF0720-5G
2-氟-4-碘甲苯 ≥95.0% (by GC)
2-氟-4-碘甲苯
分子式: C₇H₆FI
沸点: 206 °C (1013 hPa)
溶点: 22 °C
MDL编号: MFCD00001045
CAS号: 39998-81-7
EINECS: 254-740-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df9c490af0d21685d6d3e712dbef505ca5be4f7a&vid=4471526f5ecdbd8994e3b229c1e5d2f69c9ef592&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR