3-氯-2-甲基苯甲醚 ≥96.0% (by GC)

供应商: TCI

Synonyms: 2-Chloro-6-methoxytoluene, 1-Chloro-3-methoxy-2-methylbenzene

C1585-1G C1585-5G
TCIAC1585-1GEA 0 CNY
TCIAC1585-1G TCIAC1585-5G
3-氯-2-甲基苯甲醚 ≥96.0% (by GC)
3-氯-2-甲基苯甲醚
分子式: C₈H₉ClO
分子量: 156.61 g/mol
沸点: 207°C
MDL编号: MFCD00070772
CAS号: 3260-88-6
EINECS: 221-862-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6cf81bb34ef29d4b2cc7a4f5c1b183e2ada7ef04&vid=d5a553510d0f548de0fb55f434db449fe08dc62a&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR