2-Chloro-5-fluorotoluene ≥97.0% (by GC)

供应商: TCI
Warning

Synonyms: 1-Chloro-4-fluoro-2-methylbenzene

C1537-25G C1537-5G
TCIAC1537-25GEA 0 CNY
TCIAC1537-25G TCIAC1537-5G
2-Chloro-5-fluorotoluene ≥97.0% (by GC)
2-Chloro-5-fluorotoluene
分子式: C₇H₆ClF
分子量: 144.58 g/mol
沸点: 156
密度: 1.18
MDL编号: MFCD00040853
CAS号: 33406-96-1
EINECS: 251-508-6
UN号: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e238bf4cf617e745957740d7c534841a4fd4968&vid=c0a8503a30d8a29e12027b1f09baa879b769ac7a&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR