3,4-二氯三氟甲苯 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: 3,4-二氯-α,α,α-三氟甲苯

D1865-25G D1865-500G
TCIAD1865-25GEA 0 CNY
TCIAD1865-25G TCIAD1865-500G
3,4-二氯三氟甲苯 ≥98.0% (by GC)
3,4-二氯三氟甲苯
分子式: C₇H₃Cl₂F₃
分子量: 215 g/mol
沸点: 173°C
溶点: -0.2
闪点: 65°C
存储条件: Ambient
MDL编号: MFCD00000555
CAS号: 328-84-7
EINECS: 206-337-1
UN号: 1760
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2876b32127addce05f235945f51695fe997eac7&vid=72781523d1a1fec6ae04bf9f3cc25b2e18fae4bf&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR