3-甲基-2-硝基苯甲酸 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 2-硝基间甲基苯甲酸

M1370-25G M1370-250G
TCIAM1370-25GEA 0 CNY
TCIAM1370-25G TCIAM1370-250G
3-甲基-2-硝基苯甲酸 ≥98.0% (by titrimetric analysis)
3-甲基-2-硝基苯甲酸
分子式: C₈H₇NO₄
分子量: 181.15 g/mol
沸点: 340
存储条件: Ambient
MDL编号: MFCD00007180
CAS号: 5437-38-7
EINECS: 226-610-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=294f123fb9d9d56628b84e7d56e510ebc930e490&vid=5b60c8e6b6423f1d780a3e111b587b9b36b67f30&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency
Melting point 220.0 to 225.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR