3-氨基-2-甲基苯甲酸 ≥99.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: 3-氨基邻甲苯酸

A1401-25G A1401-250G
TCIAA1401-25GEA 0 CNY
TCIAA1401-25G TCIAA1401-250G
3-氨基-2-甲基苯甲酸 ≥99.0% (by HPLC, titration analysis)
3-氨基-2-甲基苯甲酸
分子式: C₈H₉NO₂
分子量: 151.16 g/mol
沸点: 347
溶点: 181
存储条件: Ambient
MDL编号: MFCD00075026
CAS号: 52130-17-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ebe6133dc7406830be5624edb8afba8bbeec14fe&vid=0808cd0205895fc14b0efccb5a2832d5706a8b46&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 186.0 to 189.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR