2-氨基-6-甲基苯甲酸 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 6-氨基邻甲苯甲酸

A0996-25G A0996-5G
TCIAA0996-25GEA 0 CNY
TCIAA0996-25G TCIAA0996-5G
2-氨基-6-甲基苯甲酸 ≥98.0% (by titrimetric analysis)
2-氨基-6-甲基苯甲酸
分子式: C₈H₉NO₂
分子量: 151.16 g/mol
沸点: 315
存储条件: Ambient
MDL编号: MFCD00007809
CAS号: 4389-50-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c39e330ecf3884f98d886bf16c59678a1b0e073&vid=d87f9044eccd34a72376e6e046e3394c94dbb22c&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Dark green powder to crystal
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR