4-叔丁基苯甲酸 ≥99.0% (by titrimetric analysis)

供应商: TCI
Danger

Synonyms: 4-(tert-Butyl)benzoic acid

B0717-25G B0717-500G
TCIAB0717-25GEA 0 CNY
TCIAB0717-25G TCIAB0717-500G
4-叔丁基苯甲酸 ≥99.0% (by titrimetric analysis)
4-叔丁基苯甲酸
分子式: C₁₁H₁₄O₂
沸点: 283 °C (1013 hPa)
溶点: 165…167 °C
密度: 0,6 g/cm³ (20 °C)
闪点: 180 °C
存储条件: Ambient
MDL编号: MFCD00002563
CAS号: 98-73-7
EINECS: 202-696-3
UN号: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f933fb4551741b713fc3f96cf29a6064c7bd9f28&vid=7b55ec18f2e8ac2a4745e3cfb44a287045f1e71d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 99.0 %
Melting point 166.0 to 168.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR