邻甲基苯甲酸 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 2-甲基苯甲酸

T0292-25G T0292-500G
TCIAT0292-25GEA 0 CNY
TCIAT0292-25G TCIAT0292-500G
邻甲基苯甲酸 ≥98.0% (by titrimetric analysis)
邻甲基苯甲酸
分子式: C₈H₈O₂
分子量: 136.15 g/mol
沸点: 259°C
闪点: 148
存储条件: Ambient
MDL编号: MFCD00002477
CAS号: 118-90-1
EINECS: 204-284-9
Merck Index: 14,09535
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44fba6356c8aa774509fd52ba72f7f9e1cd11814&vid=6756a4eceab66763be8464b58d8862ee92584bbd&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 102.0 to 107.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR