苯戊酮 ≥98.0% (by GC)

供应商: TCI

Synonyms: 丁基苯酮

V0009-25ML V0009-500ML
TCIAV0009-25MLEA 0 CNY
TCIAV0009-25ML TCIAV0009-500ML
苯戊酮 ≥98.0% (by GC)
苯戊酮
分子式: C₆H₅C(O)(CH₂)₃CH₃
分子量: 162.23 g/mol
沸点: 140°C
溶点: -9
闪点: 102°C
存储条件: Ambient
MDL编号: MFCD00009480
CAS号: 1009-14-9
EINECS: 213-767-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2584f16b192b481fbaf1d8ab1e77be6109fb67a6&vid=3bf6c22512cdad990d29bc1ffe3db3b535a55970&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Specific gravity (20/20) 0.9740 to 0.9760


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR