2-溴-6-硝基甲苯 ≥98.0% (by GC)

供应商: TCI
Warning

B1878-25G B1878-5G
TCIAB1878-25GEA 0 CNY
TCIAB1878-25G TCIAB1878-5G
2-溴-6-硝基甲苯 ≥98.0% (by GC)
2-溴-6-硝基甲苯
分子式: C₇H₆BrNO₂
沸点: 136°C
溶点: 42 °C
存储条件: Ambient
MDL编号: MFCD00009792
CAS号: 55289-35-5
EINECS: 259-566-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=325fbd3c95dbc684947228eccd5e7fad56b3fe4b&vid=c697187f6cd5ebd1aa329c7186821d62bd704b4d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White or Colorless to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Melting point 38.0 to 41.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR