2-硝基苄溴 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: α-溴-2-硝基甲苯

N0404-25G N0404-5G
TCIAN0404-25GEA 0 CNY
TCIAN0404-25G TCIAN0404-5G
2-硝基苄溴 ≥98.0% (by GC)
2-硝基苄溴
分子式: C₇H₆BrNO₂
分子量: 216.03 g/mol
存储条件: Refrigerator
MDL编号: MFCD00007184
CAS号: 3958-60-9
EINECS: 223-558-9
UN号: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=356c7441750d555a5df2ea5d652cdad641b87be1&vid=41bba3427b4c0cb48c2be8f1a3fa8e7b9974d958&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 45.0 to 48.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR