4-溴-3-(三氟甲基)苯胺 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 5-Amino-2-bromobenzotrifluoride, 4-Bromo-α,α,α-trifluoro-m-toluidine, 5-Amino-2-bromo-α,α,α-trifluorotoluene

A1847-25G A1847-5G
TCIAA1847-25GEA 0 CNY
TCIAA1847-25G TCIAA1847-5G
4-溴-3-(三氟甲基)苯胺 ≥98.0% (by GC, titration analysis)
4-溴-3-(三氟甲基)苯胺
分子式: C₇H₅BrF₃N
分子量: 240.02 g/mol
沸点: 108°C
MDL编号: MFCD00007827
CAS号: 393-36-2
EINECS: 206-885-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29f1b52869e8566096f77b8551d9ed78d680126f&vid=5e82bd17249ae54f859aeee1dc0ce28d4d073bab&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 48.0 to 53.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR