3-氨基-5-溴三氟甲苯 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 3-Amino-5-bromo-α,α,α-trifluorotoluene, 5-Bromo-α,α,α-trifluoro-m-toluidine, 3-Amino-5-bromobenzotrifluoride

A1914-25G A1914-5G
TCIAA1914-25GEA 0 CNY
TCIAA1914-25G TCIAA1914-5G
3-氨基-5-溴三氟甲苯 ≥98.0% (by GC)
3-氨基-5-溴三氟甲苯
分子式: C₇H₅BrF₃N
分子量: 240.02 g/mol
闪点: 107°C
MDL编号: MFCD00236205
CAS号: 54962-75-3
EINECS: 259-412-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b30d8bc2a2788e79169d5a16f7bc150ef5196930&vid=9adbe49dfb87450bcdfb1c2246692ff5349ec0c9&prid=web&clid=SAPDAM

现在订购

产品技术规格

Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR