2-溴-5-氟甲苯 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 1-Bromo-4-fluoro-2-methylbenzene

B1303-25G B1303-5G
TCIAB1303-25GEA 0 CNY
TCIAB1303-25G TCIAB1303-5G
2-溴-5-氟甲苯 ≥98.0% (by GC)
2-溴-5-氟甲苯
分子式: C₇H₆BrF
分子量: 189.03 g/mol
沸点: 177°C
密度: 1.49
闪点: 45°C
MDL编号: MFCD00017921
CAS号: 452-63-1
EINECS: 207-203-5
UN号: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3735fd91d6a41fe6f3bf2d3dc0545f1bf9e803b&vid=007b914c685894ebab66184cbf1fa70802791044&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR