2,5-二氟苄基溴 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: α-溴-2,5-二氟甲苯

D2597-25G D2597-5G
TCIAD2597-25GEA 0 CNY
TCIAD2597-25G TCIAD2597-5G
2,5-二氟苄基溴 ≥98.0% (by GC)
2,5-二氟苄基溴
分子式: C₇H₅BrF₂
分子量: 207.02 g/mol
沸点: 186
溶点: 8
闪点: 15°C
MDL编号: MFCD00009897
CAS号: 85117-99-3
EINECS: 285-651-0
UN号: 2924
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b07fd907811935cbdf3c01fe82ac595c091c8536&vid=602b0a71e25a71b21a23227fa87de6644ae3a377&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR