2-溴-5-氯甲苯 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
1-Bromo-4-chloro-2-methylbenzene
B1427-25G
B1427-250G
TCIAB1427-25GEA
0
CNY
TCIAB1427-25G
TCIAB1427-250G
2-溴-5-氯甲苯 ≥98.0% (by GC)
2-溴-5-氯甲苯
分子式:
C₇H₆BrCl 沸点: 100°C 密度: 1,55 g/cm³ (20 °C) 闪点: 95°C |
MDL编号:
MFCD00000602 CAS号: 14495-51-3 EINECS: 238-503-4 |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...