3-溴苄氯 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: 3-溴-α-氯甲苯

B2213-25G B2213-5G
TCIAB2213-25GEA 0 CNY
TCIAB2213-25G TCIAB2213-5G
3-溴苄氯 ≥98.0% (by GC)
3-溴苄氯
分子式: C₇H₆BrCl
分子量: 205.48 g/mol
溶点: 32.5
密度: 1.608
MDL编号: MFCD00040865
CAS号: 932-77-4
UN号: 3265
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e4c00a87508d3c0d208237d3c71a888eca102eb&vid=4164caf962f37479b40e68154d4be008165e1530&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR