3-溴-4-甲基苯甲酸 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
B3049-25G
B3049-5G
TCIAB3049-25GEA
0
CNY
TCIAB3049-25G
TCIAB3049-5G
3-溴-4-甲基苯甲酸 ≥98.0% (by GC, titration analysis)
3-溴-4-甲基苯甲酸
分子式:
C₈H₇BrO₂ 分子量: 215.05 g/mol |
MDL编号:
MFCD00002488 CAS号: 7697-26-9 EINECS: 231-712-1 |
|
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 207.0 to 213.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...