2-溴甲苯 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
B0659-25G
B0659-500G
TCIAB0659-25GEA
0
CNY
TCIAB0659-25G
TCIAB0659-500G
2-溴甲苯 ≥98.0% (by GC)
2-溴甲苯
分子式:
C₇H₇Br 分子量: 171.04 g/mol 沸点: 181°C 溶点: –26°C 闪点: 79°C 存储条件: Refrigerator |
MDL编号:
MFCD00000068 CAS号: 95-46-5 EINECS: 202-421-7 Merck Index: 13,01424 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...