3-吡啶硼酸频哪醇酯 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
3-(4,4,5,5-四甲基-1,3,2-二氧杂硼烷-2-基)吡啶, 3-吡啶基硼酸频哪醇酯
T2345-1G
T2345-5G
TCIAT2345-1GEA
0
CNY
TCIAT2345-1G
TCIAT2345-5G
3-吡啶硼酸频哪醇酯 ≥98.0% (by GC, titration analysis)
3-吡啶硼酸频哪醇酯
分子式:
C₁₁H₁₆BNO₂ 分子量: 205.06 g/mol |
MDL编号:
MFCD03084758 CAS号: 329214-79-1 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Acetone | almost transparency |
Melting point | 102.0 to 106.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...