1,4,8,11-四氮杂环十四烷 ≥98.0% (通过滴定分析)

供应商: TCI
Warning

T1597-1G T1597-5G
TCIAT1597-1GEA 0 CNY
TCIAT1597-1G TCIAT1597-5G
1,4,8,11-四氮杂环十四烷 ≥98.0% (通过滴定分析)
1,4,8,11-四氮杂环十四烷
分子式: C₁₀H₂₄N₄
MDL编号: MFCD00005105
CAS号: 295-37-4
EINECS: 206-039-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8bdc17754b7e98a4516744dfaaf07c95816f33d3&vid=8bdc17754b7e98a4516744dfaaf07c95816f33d3&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 187.0 to 193.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR