4-氨基苯硼酸频哪醇酯 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 对氨基苯硼酸频哪醇酯, 4-(4,4,5,5-四甲基-1,3,2-二氧杂环戊硼烷-2-基)苯胺

T1951-1G T1951-5G
TCIAT1951-1GEA 0 CNY
TCIAT1951-1G TCIAT1951-5G
4-氨基苯硼酸频哪醇酯 ≥98.0% (by GC)
4-氨基苯硼酸频哪醇酯
分子式: C₁₂H₁₈BNO₂
分子量: 219.09 g/mol
MDL编号: MFCD02093721
CAS号: 214360-73-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a870f77a0cfff92844b16219ee5475dc5bbcb04&vid=6a870f77a0cfff92844b16219ee5475dc5bbcb04&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 166.0 to 170.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR