2-萘硫醇 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 硫代-2-萘酚 (β), 2-萘硫酚

N0020-25G N0020-250G
TCIAN0020-25GEA 0 CNY
TCIAN0020-25G TCIAN0020-250G
2-萘硫醇 ≥98.0% (by GC, titration analysis)
2-萘硫醇
分子式: C₁₀H₈S
分子量: 160.24 g/mol
沸点: 211°C
MDL编号: MFCD00004086
CAS号: 91-60-1
EINECS: 202-082-5
Merck Index: 14,06379
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9acf4f581aeb44f3424125173c2ba08bbad281fd&vid=9acf4f581aeb44f3424125173c2ba08bbad281fd&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Iodometric Titration) min. 98.0 %
Melting point 79.0 to 83.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR