4,4'-联苯二甲酸 ≥97.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 4,4'-Biphenyldicarboxylic acid, [1,1'-Biphenyl]-4,4'-dicarboxylic acid

B1191-25G B1191-5G
TCIAB1191-25GEA 0 CNY
TCIAB1191-25G TCIAB1191-5G
4,4'-联苯二甲酸 ≥97.0% (by titrimetric analysis)
4,4'-联苯二甲酸
分子式: C₁₄H₁₀O₄
MDL编号: MFCD00002554
CAS号: 787-70-2
EINECS: 212-328-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82f79d0147604db7fcc11fcf00789f72b6f54706&vid=82f79d0147604db7fcc11fcf00789f72b6f54706&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystaline
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR