2,3-二氢呋喃 ≥98.0% (通过GC)

供应商: TCI
Danger

D1415-25ML D1415-500ML
TCIAD1415-25MLEA 0 CNY
TCIAD1415-25ML TCIAD1415-500ML
2,3-二氢呋喃 ≥98.0% (通过GC)
2,3-二氢呋喃
分子式: C₄H₆O
闪点: –25°C
MDL编号: MFCD00003205
CAS号: 1191-99-7
EINECS: 214-747-7
UN号: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a1724196c37d6e1d2ccc7f90b546ffae861d252&vid=5a1724196c37d6e1d2ccc7f90b546ffae861d252&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR