(R)-(+)-1-(1-萘基)乙胺 ≥99.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
(R)-(+)-α-甲基-1-萘甲胺
N0482-1G
N0482-5G
TCIAN0482-1GEA
0
CNY
TCIAN0482-1G
TCIAN0482-5G
(R)-(+)-1-(1-萘基)乙胺 ≥99.0% (by GC)
(R)-(+)-1-(1-萘基)乙胺
分子式:
C₁₂H₁₃N 分子量: 171.24 g/mol 沸点: 80°C 存储条件: Refrigerator |
MDL编号:
MFCD00064114 CAS号: 3886-70-2 EINECS: 223-425-5 |
|
产品技术规格
Appearance | White to Yellow to Green clear liquid |
Purity (GC) | min. 99.0 % |
Optical purity (GC) | min. 98.0 ee% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...