1-Butyl-3-methylpyridinium bromide ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: 1-Butyl-3-methylpyridin-1-ium bromide

B3104-25G B3104-5G
TCIAB3104-25GEA 0 CNY
TCIAB3104-25G TCIAB3104-5G
1-Butyl-3-methylpyridinium bromide ≥98.0% (by HPLC, titration analysis)
1-Butyl-3-methylpyridinium bromide
分子式: C₁₀H₁₆BrN
分子量: 230.15 g/mol
MDL编号: MFCD01632121
CAS号: 26576-85-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26c34fd6b8535f5d2a67a170cf25cc0cb66084c8&vid=26c34fd6b8535f5d2a67a170cf25cc0cb66084c8&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR