1-乙基-3-甲基咪唑三氟甲磺酸盐 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: EMIM Otf

E0494-25G E0494-5G
TCIAE0494-25GEA 0 CNY
TCIAE0494-25G TCIAE0494-5G
1-乙基-3-甲基咪唑三氟甲磺酸盐 ≥98.0% (by titrimetric analysis)
1-乙基-3-甲基咪唑三氟甲磺酸盐
分子式: C₇H₁₁F₃N₂O₃S
分子量: 260.24 g/mol
沸点: 671°C
溶点: –10°C
MDL编号: MFCD00210008
CAS号: 145022-44-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc49338adbdb7a363ff593c70268bb7db53d726&vid=1bc49338adbdb7a363ff593c70268bb7db53d726&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR