色胺盐酸盐 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: β-吲哚基乙胺, 3-(2-氨乙基)吲哚

A0300-1G A0300-25G
TCIAA0300-1GEA 0 CNY
TCIAA0300-1G TCIAA0300-25G
色胺盐酸盐 ≥98.0% (by titrimetric analysis)
色胺盐酸盐
分子式: C₁₀H₁₃ClN₂
溶点: 248°C
MDL编号: MFCD00012682
CAS号: 343-94-2
EINECS: 206-446-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9b7956b22879168f6e455f64b45577aba8d5211&vid=f9b7956b22879168f6e455f64b45577aba8d5211&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR