吲哚-2-羧酸乙酯 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 2-Carbethoxyinole, Ethyl 1H-indole-2-carboxylate, Indole-2-Carboxylic Acid Ethyl Ester

I0585-25G I0585-5G
TCIAI0585-25GEA 0 CNY
TCIAI0585-25G TCIAI0585-5G
吲哚-2-羧酸乙酯 ≥98.0% (by GC)
吲哚-2-羧酸乙酯
分子式: C₁₁H₁₁NO₂
分子量: 189.21 g/mol
MDL编号: MFCD00005609
CAS号: 3770-50-1
EINECS: 223-206-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd7736f5cb5c778626f2919dd461e7fa05217f18&vid=fd7736f5cb5c778626f2919dd461e7fa05217f18&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 120.0 to 125.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR