1-Boc-哌嗪 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 哌嗪-1-甲酸叔丁酯

B2415-25G B2415-5G
TCIAB2415-25GEA 0 CNY
TCIAB2415-25G TCIAB2415-5G
1-Boc-哌嗪 ≥98.0% (by GC, titration analysis)
1-Boc-哌嗪
分子式: C₉H₁₈N₂O₂
分子量: 186.25 g/mol
存储条件: Refrigerator
MDL编号: MFCD00075265
CAS号: 57260-71-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ea20ecd1b7d405cb0480ae02eb9549107cb65cf&vid=3ea20ecd1b7d405cb0480ae02eb9549107cb65cf&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 44.0 to 48.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR