1-Methyl-3-octylimidazolium bromide ≥98.0% (通过HPLC,滴定分析)
供应商: TCI
|
Warning
|
M1904-25G
M1904-5G
TCIAM1904-25GEA
0
CNY
TCIAM1904-25G
TCIAM1904-5G
1-Methyl-3-octylimidazolium bromide ≥98.0% (通过HPLC,滴定分析)
分子式:
C₁₂H₂₃BrN₂ |
MDL编号:
MFCD10566918 CAS号: 61545-99-1 |
|
产品技术规格
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...