2,6-Dibromo-4-(trifluoromethoxy)aniline ≥98.0% (by GC)
供应商: TCI
|
Warning
|
D3038-25G
D3038-5G
TCIAD3038-25GEA
0
CNY
TCIAD3038-25G
TCIAD3038-5G
2,6-Dibromo-4-(trifluoromethoxy)aniline ≥98.0% (by GC)
2,6-Dibromo-4-(trifluoromethoxy)aniline
分子式:
C₇H₄Br₂F₃NO 分子量: 334.92 g/mol |
MDL编号:
MFCD00153113 CAS号: 88149-49-9 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...