乙基丙二酸二乙酯 ≥98.0% (by GC)

供应商: TCI

E0138-25ML E0138-500ML
TCIAE0138-25MLEA 0 CNY
TCIAE0138-25ML TCIAE0138-500ML
乙基丙二酸二乙酯 ≥98.0% (by GC)
乙基丙二酸二乙酯
分子式: C₉H₁₆O₄
沸点: 211°C
闪点: 88°C
MDL编号: MFCD00009167
CAS号: 133-13-1
EINECS: 205-093-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5e6caf6d0ba9a8f3c9c6592845b5a272b6591cf&vid=f5e6caf6d0ba9a8f3c9c6592845b5a272b6591cf&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR