Methyl-4-bromobutyrate ≥98.0% (通过GC)

供应商: TCI
Warning

B2308-25G B2308-250G
TCIAB2308-25GEA 0 CNY
TCIAB2308-25G TCIAB2308-250G
Methyl-4-bromobutyrate ≥98.0% (通过GC)
Methyl-4-bromobutyrate
分子式: C₅H₉BrO₂
MDL编号: MFCD00041482
CAS号: 4897-84-1
EINECS: 225-523-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5875f00eec1563be2be0428e45714fc9ab74fb74&vid=5875f00eec1563be2be0428e45714fc9ab74fb74&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR