3,5-二甲氧基苯胺 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
5-氨基间苯二酚二甲醚
D1484-25G
D1484-5G
TCIAD1484-25GEA
0
CNY
TCIAD1484-25G
TCIAD1484-5G
3,5-二甲氧基苯胺 ≥98.0% (by GC, titration analysis)
3,5-二甲氧基苯胺
分子式:
C₈H₁₁NO₂ 分子量: 153.18 g/mol 沸点: 178°C 存储条件: Refrigerator |
MDL编号:
MFCD00008392 CAS号: 10272-07-8 EINECS: 233-616-5 |
|
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 55.0 to 58.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...