3,4-二甲氧基苯胺 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
4-Aminoveratrole, 1,2-二甲氧基-4-氨基苯
D1485-25G
D1485-500G
TCIAD1485-25GEA
0
CNY
TCIAD1485-25G
TCIAD1485-500G
3,4-二甲氧基苯胺 ≥98.0% (by GC, titration analysis)
3,4-二甲氧基苯胺
分子式:
C₈H₁₁NO₂ 分子量: 153.18 g/mol 沸点: 180°C 存储条件: Ambient |
MDL编号:
MFCD00008394 CAS号: 6315-89-5 EINECS: 228-647-6 |
|
产品技术规格
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 86.0 to 90.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...