N,N-Dimethyl-4-nitroaniline ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
D0760-25G
D0760-5G
TCIAD0760-25GEA
0
CNY
TCIAD0760-25G
TCIAD0760-5G
N,N-Dimethyl-4-nitroaniline ≥98.0% (by GC, titration analysis)
N,N-Dimethyl-4-nitroaniline
分子式:
C₈H₁₀N₂O₂ 沸点: 288 °C (1013 hPa) 溶点: 159…160 °C 密度: 1,179 g/cm³ (20 °C) 存储条件: Ambient |
MDL编号:
MFCD00014712 CAS号: 100-23-2 EINECS: 202-832-1 ADR: 6.1,III |
|
产品技术规格
Appearance | Light yellow to Brown powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 162.0 to 165.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...